N-Acetyl-L-proline 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Ac-Pro-OH
L14300.03
L14300.06
L14300.03EA
1410
CZK
L14300.03
L14300.06
N-Acetyl-L-proline 99%
N-Acetyl-L-proline
Purity: 99%
CAS No.: 68-95-1
Molecular Formula: C7H11NO3
Formula Weight: 157.18
Storage Temperature: 2°C to 8°C
Melting Point: 117-118°
Beilstein Reference: 83200
Optical Rotation: -116° (c=1 in water)
MDL No.: MFCD00020837
Vzorec:
C₇H₁₁NO₃ Bod tání: 117…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00020837 CAS číslo: 68-95-1 EINECS: 200-698-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...