Tisk…

1-(4-Bromophenyl)ethylamine 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L11848.06 L11848.03
L11848.06EA 1850 CZK
L11848.06 L11848.03
1-(4-Bromophenyl)ethylamine 96%
1-(4-Bromophenyl)ethylamine

Purity: 96% CAS No.: 24358-62-1 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Refractive Index: 1.5670 Odour: Amine-like Boiling Point: 140-145°/30mm Beilstein Reference: 2412318 UN No.: 2735 MDL No.: MFCD00025548

Vzorec: C₈H₁₀BrN
Bod varu: 140…145 °C (30 mmHg)
Hustota: 1,397
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00025548
CAS číslo: 24358-62-1
EINECS: 246-200-3
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR