Allyltriethoxysilane ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L04297.06
L04297.14
L04297.06EA
1430
CZK
L04297.06
L04297.14
Allyltriethoxysilane ≥97%
Allyltriethoxysilane
Purity: 97%
CAS No.: 2550-04-1
Molecular Formula: C9H20O3Si
Formula Weight: 204.34
Flash Point: 21°(69°F)
Refractive Index: 1.4070
Boiling Point: 176-177°
Beilstein Reference: 1772306
UN No.: 1993
MDL No.: MFCD00009064
Vzorec:
C₉H₂₀O₃Si Bod varu: 176…177 °C Hustota: 0,953 Bod vzplanutí: 21 °C (69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009064 CAS číslo: 2550-04-1 EINECS: 219-843-2 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...