Tisk…

Allyloxytrimethylsilane 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H53443.06 H53443.14
H53443.06EA 831 CZK
H53443.06 H53443.14
Allyloxytrimethylsilane 98%
Allyloxytrimethylsilane

Purity: 99% CAS No.: 18146-00-4 Molecular Formula: C6H14OSi Formula Weight: 130.26 Flash Point: 1°(34°F) Refractive Index: 1.397 Boiling Point: 101-102° Beilstein Reference: 1739517 UN No.: 1993 MDL No.: MFCD00008643

Vzorec: C₆H₁₄OSi
Bod varu: 101…102 °C
Hustota: 0,773
Bod vzplanutí: 1 °C (34 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008643
CAS číslo: 18146-00-4
EINECS: 242-031-4
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR