5-Methyl-2-thiopheneboronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Methylthiophene-2-boronic acid, (5-Methylthiophen-2-yl)boronic acid
B23138.03
B23138.06
B23138.03EA
1460
CZK
B23138.03
B23138.06
5-Methyl-2-thiopheneboronic acid ≥98%
5-Methyl-2-thiopheneboronic acid
Purity: 98%
CAS No.: 162607-20-7
Molecular Formula: C5H7BO2S
Formula Weight: 142.01
Storage Temperature: 2°C to 8°C
Melting Point: ca 160° dec.
Beilstein Reference: 7019569
MDL No.: MFCD01318166
Vzorec:
C₅H₇BO₂S MW: 141,99 g/mol Bod tání: ∼ 160 °C (rozkl.) |
MDL Number:
MFCD01318166 CAS číslo: 162607-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...