Tisk…

β-Chloro-4'-fluoropropiophenone ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone

B21891.06 B21891.14 B21891.22
B21891.06EA 889 CZK
B21891.06 B21891.14 B21891.22
β-Chloro-4'-fluoropropiophenone ≥97%
β-Chloro-4'-fluoropropiophenone

Purity: 97% CAS No.: 347-93-3 Molecular Formula: C9H8ClFO Formula Weight: 186.61 Melting Point: 46-49° Boiling Point: 100-103°/3mm MDL No.: MFCD00000991

Vzorec: C₉H₈ClFO
MW: 186,61 g/mol
Bod varu: 100…103 °C (3 mmHg)
Bod tání: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00000991
CAS číslo: 347-93-3
EINECS: 206-475-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR