N-(2-Aminoethyl)morpholine ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-(2-Aminoethyl)morpholine, 2-Morpholinoethanamine
Purity: 98+%
CAS No.: 2038-03-1
Molecular Formula: C6H14N2O
Formula Weight: 130.19
Flash Point: 86°(186°F)
Refractive Index: 1.4760
Odour: Amine-like
pH: 12
Melting Point: 23-25°
Boiling Point: 200-203°
Beilstein Reference: 104378
UN No.: 2923
MDL No.: MFCD00006182
Vzorec:
C₆H₁₄N₂O MW: 130,19 g/mol Bod varu: 200…203 °C Bod tání: 23…25 °C Hustota: 0,999 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006182 CAS číslo: 2038-03-1 EINECS: 218-011-6 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...