3-Ethoxy-2-thenoic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B20296.03
B20296.06
B20296.14
B20296.03EA
2300
CZK
B20296.03
B20296.06
B20296.14
3-Ethoxy-2-thenoic acid ≥97%
3-Ethoxy-2-thenoic acid
Purity: 97%
CAS No.: 139926-23-1
Molecular Formula: C7H8O3S
Formula Weight: 172.20
Melting Point: 121-123°
MDL No.: MFCD00205205
Vzorec:
C₇H₈O₃S Bod tání: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00205205 CAS číslo: 139926-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...