Tisk…

4-Butoxyaniline ≥97%

Dodavatel: Thermo Scientific
Varování

A17528.06 A17528.14
A17528.06EA 1880 CZK
A17528.06 A17528.14
4-Butoxyaniline ≥97%
4-Butoxyaniline

Purity: 97% CAS No.: 4344-55-2 Molecular Formula: C10H15NO Formula Weight: 165.24 Flash Point: 110°(230°F) Refractive Index: 1.5380 Boiling Point: 84-86°/0.05mm Beilstein Reference: 2084421 UN No.: 2810 MDL No.: MFCD00007866

Vzorec: C₁₀H₁₅NO
MW: 165,24 g/mol
Bod varu: 84…86 °C (0,05 mmHg)
Hustota: 0,992
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007866
CAS číslo: 4344-55-2
EINECS: 224-402-2
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR