Cyclooctanone oxime ≥98%
Dodavatel: Thermo Scientific
A16672.09
A16672.18
A16672.09EA
1880
CZK
A16672.09
A16672.18
Cyclooctanone oxime ≥98%
Cyclooctanone oxime
Purity: 98+%
CAS No.: 1074-51-7
Molecular Formula: C8H15NO
Formula Weight: 141.22
Melting Point: 36-37°
Boiling Point: 128-129°/14mm
MDL No.: MFCD00191967
Vzorec:
C₈H₁₅NO Bod varu: 128…129 °C (14 mmHg) Bod tání: 36…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191967 CAS číslo: 1074-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...