1-(Trimethylsiloxy)cyclohexene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(Cyclohex-1-en-1-yloxy)trimethylsilane
A16099.09
A16099.18
A16099.09EA
2020
CZK
A16099.09
A16099.18
1-(Trimethylsiloxy)cyclohexene ≥98%
1-(Trimethylsiloxy)cyclohexene
Purity: 98%
CAS No.: 6651-36-1
Molecular Formula: C9H18OSi
Formula Weight: 170.33
Flash Point: 42°(107°F)
Refractive Index: 1.4470
Boiling Point: 165°
Beilstein Reference: 1859394
UN No.: 1993
MDL No.: MFCD00001541
Vzorec:
C₉H₁₈OSi MW: 170.33 Bod varu: 165 °C Hustota: 0.883 Bod vzplanutí: 42 °C (107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001541 CAS číslo: 6651-36-1 EINECS: 229-675-1 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...