4-Nitrophenethyl bromide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene
A15677.06
A15677.14
A15677.06EA
2000
CZK
A15677.06
A15677.14
4-Nitrophenethyl bromide ≥98%
4-Nitrophenethyl bromide
Purity: 98%
CAS No.: 5339-26-4
Molecular Formula: C8H8BrNO2
Formula Weight: 230.07
Melting Point: 67-71°
Boiling Point: 116°/0.2mm
MDL No.: MFCD00007386
Vzorec:
C₈H₈BrNO₂ MW: 230.06 Bod varu: 116 °C (0.2 mmHg) Bod tání: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007386 CAS číslo: 5339-26-4 EINECS: 226-271-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...