Tisk…

4-Chloro-7-nitrobenzofurazan ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: NBD chloride, 4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, NBD-Cl

A14165.06 A14165.14
A14165.06EA 2220 CZK
A14165.06 A14165.14
4-Chloro-7-nitrobenzofurazan ≥99%
4-Chloro-7-nitrobenzofurazan

Purity: 99% CAS No.: 10199-89-0 Molecular Formula: C6H2ClN3O3 Formula Weight: 199.56 Melting Point: 96-100° Beilstein Reference: 614212

Vzorec: C₆H₂ClN₃O₃
MW: 199,55 g/mol
Bod tání: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00005808
CAS číslo: 10199-89-0
EINECS: 233-496-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR