Tisk…

Dichloro(diphenyl)silane ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12051.36 A12051.22
A12051.36EA 5120 CZK
A12051.36 A12051.22
Dichloro(diphenyl)silane ≥97%
Dichloro(diphenyl)silane

Purity: 97% CAS No.: 80-10-4 Molecular Formula: C12H10Cl2Si Formula Weight: 253.20 Flash Point: 157°(314°F) Refractive Index: 1.5790 Odour: Pungent Melting Point: -22° Boiling Point: 303-304° Beilstein Reference: 609882 UN No.: 1769 MDL No.: MFCD00000489

Vzorec: C₁₂H₁₀Cl₂Si
Bod varu: 303…304 °C
Bod tání: –22 °C
Hustota: 1,208
Bod vzplanutí: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000489
CAS číslo: 80-10-4
EINECS: 201-251-0
UN: 1769
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR