Benzoin methyl ether ≥97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone
A10296.14
A10296.22
A10296.36
A10296.14EA
1650
CZK
A10296.14
A10296.22
A10296.36
Benzoin methyl ether ≥97%
Benzoin methyl ether
Purity: 97%
CAS No.: 3524-62-7
Molecular Formula: C15H14O2
Formula Weight: 226.28
Flash Point:
Vzorec:
C₁₅H₁₄O₂ MW: 226.27 Bod varu: 188…189 °C (15 mmHg) Bod tání: 46…50 °C Hustota: 1.128 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008492 CAS číslo: 3524-62-7 EINECS: 222-538-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...