Tisk…

Benzoin ≥99%

Dodavatel: Thermo Scientific

Synonyms: 2-Hydroxy-2-phenylacetophenone

A10188.0B A10188.0I A10188.30
A10188.0BEA 2580 CZK
A10188.0B A10188.0I A10188.30
Benzoin ≥99%
Benzoin

Purity: 99% CAS No.: 119-53-9 Molecular Formula: C14H12O2 Formula Weight: 212.25 Flash Point: 181°(357°F) Odour: Pleasant Boiling Point: 334-344° Beilstein Reference: 391839 Merck Reference: 14,1093 MDL No.: MFCD00004496

Vzorec: C₁₄H₁₂O₂
MW: 212,25 g/mol
Bod varu: 334…344 °C
Bod tání: 131…136 °C
Hustota: 1,310
Bod vzplanutí: 181 °C (357 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004496
CAS číslo: 119-53-9
EINECS: 204-331-3
Merck Index: 14,1093

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR