Tisk…

2,6-Dimethylbenzonitrile ≥97%

Dodavatel: Thermo Scientific
Varování

A14461.03 A14461.06 A14461.14
A14461.03EA 876 CZK
A14461.03 A14461.06 A14461.14
2,6-Dimethylbenzonitrile ≥97%
2,6-Dimethylbenzonitrile

Purity: 97% CAS No.: 6575-13-9 Molecular Formula: C9H9N Formula Weight: 131.18 Odour: Almond-like Melting Point: 88-92° Beilstein Reference: 1932523 MDL No.: MFCD00037166

Vzorec: C₉H₉N
MW: 131,18 g/mol
Bod tání: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00037166
CAS číslo: 6575-13-9
EINECS: 229-503-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR