2-Methoxypropene 97% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene
174350010
174352500
174350025
ACRO174350010EA
7620
CZK
ACRO174350010
ACRO174352500
ACRO174350025
2-Methoxypropene 97% stabilised
2-Methoxypropene
Vzorec:
C₄H₈O MW: 72.11 Hustota: 0.78 g/cm³ Bod vzplanutí: –29 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014929 CAS číslo: 116-11-0 EINECS: 204-125-3 UN: 1993 ADR: 3 I |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3810 to 1.3840 (20°C, 589 nm) |
GC | ≥96.0 % |
Stabilizer | ≤0.5 % potassium carbonate |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...