Dichloro(diphenyl)silane 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Diphenyldichlorosilane, Dichlorodiphenylsilane
113341000
113345000
ACRO113341000EA
1630
CZK
ACRO113341000
ACRO113345000
Dichloro(diphenyl)silane 97%
Dichloro(diphenyl)silane
Vzorec:
C₁₂H₁₀Cl₂Si MW: 253.2 Bod varu: 305 °C (760 mmHg) Bod tání: –22 °C Hustota: 1.204 g/cm³ Bod vzplanutí: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000489 CAS číslo: 80-10-4 EINECS: 201-251-0 UN: 1769 ADR: 8 II |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5765 to 1.5815 (20°C, 589 nm) |
GC | ≥96.0 % |
Absorption Ratio P-15/P+15 | 1.1 to 1.32 |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...