Tisk…

3,5-Di-tert-butyl-4-hydroxybenzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A15864.06 A15864.14 A15864.22 AB109337.0025
A15864.06EA 1060 CZK
A15864.06 A15864.14 A15864.22 ABCRAB109337.0025
3,5-Di-tert-butyl-4-hydroxybenzaldehyde ≥98%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Purity: 98+% CAS No.: 1620-98-0 Molecular Formula: C15H22O2 Formula Weight: 234.34 Melting Point: 187-189° Beilstein Reference: 982526

Vzorec: C₁₅H₂₂O₂
MW: 234.34
Bod varu: 332
Bod tání: 187…189 °C
Hustota: 1.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008826
CAS číslo: 1620-98-0
EINECS: 216-592-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR