Tisk…

α'-Bromo-α,α,α-trifluoro-m-xylene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethyl)benzene, 3-(Bromomethyl)benzotrifluoride

A19330.06 A19330.14
A19330.06EA 817 CZK
A19330.06 A19330.14
α'-Bromo-α,α,α-trifluoro-m-xylene ≥98%
α'-Bromo-α,α,α-trifluoro-m-xylene

Purity: 98% CAS No.: 402-23-3 Molecular Formula: C8H6BrF3 Formula Weight: 239.04 Flash Point: 87°(188°F) Refractive Index: 1.4920 Odour: Pungent Boiling Point: 67-69°/4mm Beilstein Reference: 743243 UN No.: 3265 MDL No.: MFCD00000395

Vzorec: C₈H₆BrF₃
Bod varu: 67…69 °C (4 mmHg)
Hustota: 1,565
Bod vzplanutí: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000395
CAS číslo: 402-23-3
EINECS: 206-938-9
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR