Tisk…

2-Bromo-m-xylene ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Bromo-1,3-dimethylbenzene, 2,6-Dimethylbromobenzene

A13130.09 A13130.14 A13130.22 A13130.30 A13130.36
A13130.09EA 1220 CZK
A13130.09 A13130.14 A13130.22 A13130.30 A13130.36
2-Bromo-m-xylene ≥98%
2-Bromo-m-xylene

Purity: 98+% CAS No.: 576-22-7 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 73°(163°F) Refractive Index: 1.5550 Melting Point: -10° Boiling Point: 205-206° Beilstein Reference: 1929780 MDL No.: MFCD00000075

Vzorec: C₈H₉Br
MW: 185.06
Bod varu: 205…206 °C
Bod tání: –10 °C
Hustota: 1.384
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000075
CAS číslo: 576-22-7
EINECS: 209-397-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR