α,α'-Dibromo-p-xylene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
p-Xylylene dibromide, 1,4-Bis(bromomethyl)benzene
M-Clarity™ quality level = MQ100
34440-100G-F
34440-25G-F
SIAL34440-100G-FEA
8220
CZK
SIAL34440-100G-F
SIAL34440-25G-F
α,α'-Dibromo-p-xylene, Sigma-Aldrich®
α,α'-Dibromo-p-xylene
Vzorec:
C₈H₈Br₂ MW: 263.96 Bod varu: 245 Hustota: 2.012 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000182 CAS číslo: 623-24-5 EINECS: 210-781-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...