α,α,α',α'-Tetrabromo-o-xylene ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1,2-Bis(dibromomethyl)benzene
A14121.30
A14121.18
A14121.30EA
5770
CZK
A14121.30
A14121.18
α,α,α',α'-Tetrabromo-o-xylene ≥97%
α,α,α',α'-Tetrabromo-o-xylene
Purity: 97%
CAS No.: 13209-15-9
Molecular Formula: C8H6Br4
Formula Weight: 421.75
Melting Point: 114-118°
Beilstein Reference: 2211698
UN No.: 1759
MDL No.: MFCD00000131
Vzorec:
C₈H₆Br₄ MW: 421,75 g/mol Bod varu: 346 °C (1013 hPa) Bod tání: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000131 CAS číslo: 13209-15-9 EINECS: 236-176-2 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...