Tisk…

tritylchlorid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane

M-Clarity™ quality level = MQ200

93000-100G 93000-500G 93000-5KG
SIAL93000-100GEA 1810 CZK
SIAL93000-100G SIAL93000-500G SIAL93000-5KG
tritylchlorid, Sigma-Aldrich®
tritylchlorid
Vzorec: C₁₉H₁₅Cl
MW: 278,78 g/mol
Bod varu: 374 °C (1013 hPa)
Bod tání: 109…112 °C
Hustota: 1,25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS číslo: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR