4-Phenylbutyrophenone ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
1,4-Diphenyl-1-butanone
L15597.03
L15597.06
L15597.14
L15597.03EA
1210
CZK
L15597.03
L15597.06
L15597.14
4-Phenylbutyrophenone ≥98%
4-Phenylbutyrophenone
Purity: 98%
CAS No.: 5407-91-0
Molecular Formula: C16H16O
Formula Weight: 224.30
Melting Point: 54-56°
Boiling Point: 180°/8mm
MDL No.: MFCD00022005
Vzorec:
(C₆H₅)C(O)CH₂CH₂CH₂(C₆H₅) Bod varu: 180 °C (8 mmHg) Bod tání: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022005 CAS číslo: 5407-91-0 EINECS: 226-471-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...