Tisk…

Propylbenzen ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B21468.22 B21468.36
B21468.22EA 1610 CZK
B21468.22 B21468.36
Propylbenzen ≥98%
Propylbenzen

Purity: 98% CAS No.: 103-65-1 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 47°(116°F) Refractive Index: 1.4910 Melting Point: -100° Boiling Point: 159-160° Beilstein Reference: 1903006 Merck Reference: 14,7844 UN No.: 1993 MDL No.: MFCD00009377

Vzorec: C₉H₁₂
Bod varu: 159…160 °C
Bod tání: –100 °C
Hustota: 0,862
Bod vzplanutí: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009377
CAS číslo: 103-65-1
EINECS: 203-132-9
UN: 1993
ADR: 3,III
Merck Index: 14,7844

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR