Tisk…
Non return valves
Dodavatel: BURKLE
8714-0100 8714-0060 8714-0080 8714-1008 8714-1006 8714-1010
229-0136EA 109 CZK
229-0136 229-0134 229-0135 229-0137 BURK8714-1006 BURK8714-1010
Non return valves
Ventily Ventily nevratné
Translucent PP retaining valves with embossed directional arrow or printed in black. With tapered ends. Various uses.
  • Resistant to compression max. 4 bar
  • Operating temperature up to +40 °C
  • With PP ball valve