n-Hexylbenzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Phenylhexane, Hexylbenzene
A12821.14
A12821.22
A12821.36
A12821.14EA
2140
CZK
A12821.14
A12821.22
A12821.36
n-Hexylbenzene ≥98%
n-Hexylbenzene
Purity: 98%
CAS No.: 1077-16-3
Molecular Formula: C12H18
Formula Weight: 162.28
Flash Point: 83°(181°F)
Refractive Index: 1.4860
Melting Point: -61°
Boiling Point: 226-227°
Beilstein Reference: 1904786
UN No.: 3082
MDL No.: MFCD00009526
Vzorec:
C₁₂H₁₈ MW: 162,27 g/mol Bod varu: 226…227 °C Bod tání: –61 °C Hustota: 0,861 Bod vzplanutí: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009526 CAS číslo: 1077-16-3 EINECS: 214-070-7 UN: 3082 ADR: 9,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...