ethylbenzen ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phenylethane
L05908.AE
L05908.AP
L05908.0F
L05908.AEEA
855
CZK
L05908.AE
L05908.AP
L05908.0F
ethylbenzen ≥99%
ethylbenzen
Purity: 99%
CAS No.: 100-41-4
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 15°(59°F)
Refractive Index: 1.4950
Odour: Petrol-like
Melting Point: -95°
Boiling Point: 135-136°
Beilstein Reference: 1901871
Merck Reference: 14,3765
UN No.: 1175
MDL No.: MFCD00011647
Vzorec:
C₈H₁₀ Bod varu: 135…136 °C Bod tání: –95 °C Hustota: 0,867 Bod vzplanutí: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011647 CAS číslo: 100-41-4 EINECS: 202-849-4 UN: 1175 ADR: 3,II Merck Index: 14,3765 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...