Butylbenzene ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
n-Butylbenzene
A10910.0B
A10910.18
A10910.30
A10910.0BEA
14620
CZK
A10910.0B
A10910.18
A10910.30
Butylbenzene ≥99%
Butylbenzene
Purity: 99%
CAS No.: 104-51-8
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 59°(138°F)
Refractive Index: 1.4890
Odour: Aromatic
Melting Point: -88°
Boiling Point: 181-183°
Beilstein Reference: 1903395
Merck Reference: 14,1549
UN No.: 2709
MDL No.: MFCD00009463
Vzorec:
C₁₀H₁₄ MW: 134,22 g/mol Bod varu: 181…183 °C Bod tání: –88 °C Hustota: 0,860 Bod vzplanutí: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009463 CAS číslo: 104-51-8 EINECS: 203-209-7 UN: 2709 ADR: 3,III Merck Index: 14,1549 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...