4-Pentylphenol ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Amylphenol
A11514.06
A11514.14
A11514.06EA
1540
CZK
A11514.06
A11514.14
4-Pentylphenol ≥98%
4-Pentylphenol
Purity: 98%
CAS No.: 14938-35-3
Molecular Formula: C11H16O
Formula Weight: 164.25
Flash Point: 133°(271°F)
Refractive Index: 1.5145
Melting Point: 24°
Boiling Point: 342°
Beilstein Reference: 1364424
UN No.: 2430
MDL No.: MFCD00020211
Vzorec:
C₁₁H₁₆O MW: 164,25 g/mol Bod varu: 342 °C Bod tání: 24 °C Hustota: 0,960 Bod vzplanutí: 133 °C (271 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020211 CAS číslo: 14938-35-3 EINECS: 239-015-4 UN: 2430 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...