Tisk…

2-sec-Butylphenol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A18461.0B A18461.30
A18461.0BEA 2670 CZK
A18461.0B A18461.30
2-sec-Butylphenol ≥98%
2-sec-Butylphenol

Purity: 98% CAS No.: 89-72-5 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 112°(233°F) Refractive Index: 1.5220 Odour: Phenol-like Melting Point: 10-12° Boiling Point: 226-228° Beilstein Reference: 1210026 UN No.: 3145 MDL No.: MFCD00002225

Vzorec: C₁₀H₁₄O
Bod varu: 226…228 °C
Bod tání: 10…12 °C
Hustota: 0,982
Bod vzplanutí: 112 °C (233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002225
CAS číslo: 89-72-5
EINECS: 201-933-8
UN: 3145
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR