3-Ethylphenol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
04688-100ML
04688-25ML
SIAL04688-100MLEA
9040
CZK
SIAL04688-100ML
SIAL04688-25ML
3-Ethylphenol, Sigma-Aldrich®
3-Ethylphenol
Vzorec:
C₈H₁₀O Bod varu: 212 °C (1013 hPa) Bod tání: –4 °C Hustota: 1,0283 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002311 CAS číslo: 620-17-7 EINECS: 210-627-3 UN: 3145 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...