Tisk…

3-tert-Butylphenol ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: m-tert-Butylphenol

B24315.06 B24315.14 B24315.22
B24315.06EA 2070 CZK
B24315.06 B24315.14 B24315.22
3-tert-Butylphenol ≥99%
3-tert-Butylphenol

Purity: 99% CAS No.: 585-34-2 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 108°(228°F) Melting Point: 44-46° Boiling Point: 125-130°/20mm UN No.: 2430 MDL No.: MFCD00002300

Vzorec: C₁₀H₁₄O
MW: 150,22 g/mol
Bod varu: 125…130 °C (20 mmHg)
Bod tání: 44…46 °C
Hustota: 1,048 g/cm³ (20 °C)
Bod vzplanutí: 108 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002300
CAS číslo: 585-34-2
EINECS: 209-553-4
UN: 2430
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR