2-tert-Butylphenol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L07294.22
L07294.36
L07294.0P
L07294.22EA
855
CZK
L07294.22
L07294.36
L07294.0P
2-tert-Butylphenol ≥99%
2-tert-Butylphenol
Purity: 99%
CAS No.: 88-18-6
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 102°(215°F)
Refractive Index: 1.5230
Melting Point: -7°
Boiling Point: 223-224°
Beilstein Reference: 1907120
UN No.: 2922
MDL No.: MFCD00002223
Vzorec:
C₁₀H₁₄O MW: 150.22 Bod varu: 223…224 °C Bod tání: –7 °C Hustota: 0.980 Bod vzplanutí: 102 °C (215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002223 CAS číslo: 88-18-6 EINECS: 201-807-2 UN: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...