Tisk…

4-Methylveratrole ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1,2-Dimethoxy-4-methylbenzene, 3,4-Dimethoxytoluene

A15558.06 A15558.14
A15558.06EA 1650 CZK
A15558.06 A15558.14
4-Methylveratrole ≥98%
4-Methylveratrole

Purity: 98% CAS No.: 494-99-5 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 84°(183°F) Refractive Index: 1.5280 Odour: Aromatic Melting Point: 25-28° Boiling Point: 220° Beilstein Reference: 2045328 MDL No.: MFCD00016651

Vzorec: C₉H₁₂O₂
MW: 152.19
Bod varu: 220 °C
Bod tání: 25…28 °C
Hustota: 1.059
Bod vzplanutí: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016651
CAS číslo: 494-99-5
EINECS: 207-796-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR