Tisk…

4-Isopropylanisole 95%

Dodavatel: Thermo Scientific
Varování

A16970.14 A16970.22
A16970.14EA 2190 CZK
A16970.14 A16970.22
4-Isopropylanisole 95%
4-Isopropylanisole

Purity: 95% CAS No.: 4132-48-3 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 83°(181°F) Refractive Index: 1.5030 Boiling Point: 212-213° Beilstein Reference: 2080269 MDL No.: MFCD00015033

Vzorec: C₁₀H₁₄O
MW: 150,22 g/mol
Bod varu: 212…213 °C
Hustota: 0,935
Bod vzplanutí: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015033
CAS číslo: 4132-48-3
EINECS: 223-952-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR