1-Nitro-4-propylbenzene 96%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L07730.06
L07730.14
L07730.06EA
3570
CZK
L07730.06
L07730.14
1-Nitro-4-propylbenzene 96%
1-Nitro-4-propylbenzene
Purity: 96%
CAS No.: 10342-59-3
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Refractive Index: 1.5370
Melting Point: -14°
Boiling Point: 96-100°/2mm
Beilstein Reference: 2043939
UN No.: 2810
MDL No.: MFCD00051839
Vzorec:
C₉H₁₁NO₂ Bod varu: 96…100 °C (2 mmHg) Bod tání: –14 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051839 CAS číslo: 10342-59-3 EINECS: 233-743-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...