Tisk…

2-Nitrotoluen ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A13446.0E A13446.22 A13446.36
A13446.0EEA 2280 CZK
A13446.0E A13446.22 A13446.36
2-Nitrotoluen ≥99%
2-Nitrotoluen

Purity: 99+% CAS No.: 88-72-2 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Flash Point: 106°(222°F) Refractive Index: 1.5450 Melting Point: -4° Boiling Point: 224-226° Beilstein Reference: 1907580 Merck Reference: 14,6650 UN No.: 1664 MDL No.: MFCD00007157

Vzorec: C₇H₇NO₂
MW: 137,14 g/mol
Bod varu: 224…226 °C
Bod tání: –4 °C
Hustota: 1,166
Bod vzplanutí: 106 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007157
CAS číslo: 88-72-2
EINECS: 201-853-3
UN: 1664
ADR: 6.1,II
Merck Index: 14,6650

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR