Tisk…

4-tert-Butylaniline ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-(tert-Butyl)aniline

A10925.06 A10925.14 A10925.22
A10925.06EA 2040 CZK
A10925.06 A10925.14 A10925.22
4-tert-Butylaniline ≥99%
4-tert-Butylaniline

Purity: 99% CAS No.: 769-92-6 Molecular Formula: C10H15N Formula Weight: 149.23 Flash Point: 101°(213°F) Refractive Index: 1.5380 Melting Point: 15-16° Boiling Point: 235° Beilstein Reference: 2205786 MDL No.: MFCD00007899

Vzorec: C₁₀H₁₅N
MW: 149,24 g/mol
Bod varu: 235 °C
Bod tání: 15…16 °C
Hustota: 0,941
Bod vzplanutí: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007899
CAS číslo: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR