Tisk…

3-tert-Butylaniline ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-(tert-Butyl)aniline, m-tert-Butylaniline

H50698.03 H50698.06
H50698.03EA 5560 CZK
H50698.03 H50698.06
3-tert-Butylaniline ≥97%
3-tert-Butylaniline

Purity: 97% CAS No.: 5369-19-7 Molecular Formula: C10H15N Formula Weight: 149.23 MDL No.: MFCD00125078

Vzorec: C₁₀H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD00125078
CAS číslo: 5369-19-7
EINECS: 226-361-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR