4-Iodotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A15631.14
A15631.22
A15631.36
A15631.14EA
1170
CZK
A15631.14
A15631.22
A15631.36
4-Iodotoluene ≥98%
4-Iodotoluene
Purity: 98%
CAS No.: 624-31-7
Molecular Formula: C7H7I
Formula Weight: 218.04
Flash Point: 90°(194°F)
Melting Point: 33-35°
Boiling Point: 210-212°
Beilstein Reference: 1903637
MDL No.: MFCD00001059
Vzorec:
C₇H₇I Bod varu: 210…212 °C Bod tání: 33…35 °C Hustota: 1,678 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001059 CAS číslo: 624-31-7 EINECS: 210-841-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...