Tisk…

2-Iodotoluene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Iodo-2-methylbenzene

B22697.14 B22697.22 B22697.36
B22697.14EA 1350 CZK
B22697.14 B22697.22 B22697.36
2-Iodotoluene ≥98%
2-Iodotoluene

Purity: 98% CAS No.: 615-37-2 Molecular Formula: C7H7I Formula Weight: 218.04 Flash Point: 90°(194°F) Refractive Index: 1.6090 Boiling Point: 210-212° Beilstein Reference: 1634294 MDL No.: MFCD00001042

Vzorec: C₇H₇I
MW: 218,04 g/mol
Bod varu: 210…212 °C
Hustota: 1,713
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001042
CAS číslo: 615-37-2
EINECS: 210-422-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR