4-Nitro-α,α,α-trifluorotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Nitro-alpha,alpha,alpha-trifluorotoluene, 4-Nitrobenzotrifluoride, 1-Nitro-4-(trifluoromethyl)benzene
L15766.03
L15766.06
L15766.03EA
972
CZK
L15766.03
L15766.06
4-Nitro-α,α,α-trifluorotoluene ≥98%
4-Nitro-α,α,α-trifluorotoluene
Purity: 98%
CAS No.: 402-54-0
Molecular Formula: C7H4F3NO2
Formula Weight: 191.11
Flash Point: 88°(190°F)
Odour: Characteristic
Boiling Point: 81-83°/10mm
UN No.: 3431
MDL No.: MFCD00007358
Vzorec:
C₇H₄F₃NO₂ Bod varu: 81…83 °C (10 mmHg) Bod tání: 38…42 °C Bod vzplanutí: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007358 CAS číslo: 402-54-0 EINECS: 206-948-3 UN: 3431 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...