Tisk…

3-Fluoro-4-nitrotoluene ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10840.06 A10840.14
A10840.06EA 996 CZK
A10840.06 A10840.14
3-Fluoro-4-nitrotoluene ≥99%
3-Fluoro-4-nitrotoluene

Purity: 99% CAS No.: 446-34-4 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Flash Point: 110°(230°F) Melting Point: 51-55° Boiling Point: 97-98°/3mm Beilstein Reference: 2502584 UN No.: 2811 MDL No.: MFCD00007053

Vzorec: C₇H₆FNO₂
MW: 155.13
Bod varu: 97…98 °C (3 mmHg)
Bod tání: 51…55 °C
Hustota: 1.438
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007053
CAS číslo: 446-34-4
EINECS: 207-166-5
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR