Tisk…

(4-Fluorophenyl)acetone ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Fluorophenylacetone

L11720.03 L11720.06
L11720.03EA 1050 CZK
L11720.03 L11720.06
(4-Fluorophenyl)acetone ≥99%
(4-Fluorophenyl)acetone

Purity: 99% CAS No.: 459-03-0 Molecular Formula: C9H9FO Formula Weight: 152.17 Flash Point: 91°(195°F) Refractive Index: 1.4958 Boiling Point: 106-107°/18mm Beilstein Reference: 1936697 MDL No.: MFCD00000362

Vzorec: C₉H₉FO
MW: 152,17 g/mol
Bod varu: 106…107 °C (18 mmHg)
Hustota: 1,11
Bod vzplanutí: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000362
CAS číslo: 459-03-0
EINECS: 207-284-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR