Perfluorotoluene ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Octafluorotoluene, Pentafluoro(trifluoromethyl)benzene, 1,2,3,4,5-Pentafluoro-6-(trifluoromethyl)benzene, α,α,α,2,3,4,5,6-Octafluorotoluene
Purity: 98+%
CAS No.: 434-64-0
Molecular Formula: C7F8
Formula Weight: 236.06
Flash Point: 20°(68°F)
Refractive Index: 1.3680
Melting Point: -66°
Boiling Point: 103-105°
Beilstein Reference: 2057072
UN No.: 1992
MDL No.: MFCD00000375
Vzorec:
C₇F₈ MW: 236,06 g/mol Bod varu: 103…105 °C Bod tání: –66 °C Hustota: 1,668 Bod vzplanutí: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000375 CAS číslo: 434-64-0 EINECS: 207-104-7 UN: 1992 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...