α,α,α,3-Tetrafluorotoluene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Fluorobenzotrifluoride, 1-Fluoro-3-(trifluoromethyl)benzene
A15055.14
A15055.22
A15055.14EA
1470
CZK
A15055.14
A15055.22
α,α,α,3-Tetrafluorotoluene ≥99%
α,α,α,3-Tetrafluorotoluene
Purity: 99%
CAS No.: 401-80-9
Molecular Formula: C7H4F4
Formula Weight: 164.10
Flash Point: 7°(44°F)
Refractive Index: 1.4010
Boiling Point: 101-102°
Beilstein Reference: 2045664
UN No.: 1993
MDL No.: MFCD00000382
Vzorec:
C₇H₄F₄ MW: 164,1 g/mol Bod varu: 101…102 °C Bod tání: –81,5 °C Hustota: 1,303 Bod vzplanutí: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000382 CAS číslo: 401-80-9 EINECS: 206-933-1 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...