Tisk…

2-Fluorotoluene ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Fluoro-2-methylbenzene, o-Fluorotoluene

A16912.0E A16912.22 A16912.36
A16912.0EEA 14900 CZK
A16912.0E A16912.22 A16912.36
2-Fluorotoluene ≥99%
2-Fluorotoluene

Purity: 99+% CAS No.: 95-52-3 Molecular Formula: C7H7F Formula Weight: 110.13 Flash Point: 12°(53°F) Refractive Index: 1.4730 Odour: Aromatic Melting Point: -87° Boiling Point: 113-114° Beilstein Reference: 1853362 Merck Reference: 14,4180 UN No.: 2388 MDL No.: MFCD00000322

Vzorec: C₇H₇F
MW: 110,13 g/mol
Bod varu: 113…114 °C
Bod tání: –87 °C
Hustota: 1,002
Bod vzplanutí: 12 °C (53 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000322
CAS číslo: 95-52-3
EINECS: 202-428-5
UN: 2388
ADR: 3,II
Merck Index: 14,4180

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR